Assay Details
| Gene Symbol: birc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145195.1 | NP_660196.1 | 2 - 3 | 221 | 102 | |
| XM_021469721.1 | XP_021325396.1 | 2 - 3 | 249 | 102 | ||
| GenBank mRNA | AY057058.1 | - | 2 - 3 | 221 | 102 | |
| BC105963.1 | - | 2 - 3 | 274 | 102 | ||
More Information
Set Membership: |
|

birc5b
Genomic Map