Assay Details
| Gene Symbol: appb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152886.1 | NP_690842.1 | 1 - 2 | 162 | 66 | |
| GenBank mRNA | AJ315639.1 | - | 1 - 2 | 162 | 66 | |
| BC083534.1 | - | 1 - 2 | 179 | 66 | ||
| CU638798.1 | - | 1 - 2 | 74 | 66 | ||
More Information
Set Membership: |
|

appb
Genomic Map