Assay Details
| Gene Symbol: ptgs2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153657.1 | NP_705943.1 | 2 - 3 | 250 | 72 | |
| GenBank mRNA | AY028585.1 | - | 2 - 3 | 250 | 72 | |
| BC056736.1 | - | 2 - 3 | 239 | 72 | ||
| BC063232.1 | - | 2 - 3 | 225 | 72 | ||
More Information
Set Membership: |
|

ptgs2a
Genomic Map