Assay Details
Gene Symbol: ncbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173249.1 | NP_775356.1 | 3 - 4 | 450 | 77 | |
GenBank mRNA | AY099530.1 | - | 3 - 4 | 450 | 77 | |
BC083529.1 | - | 3 - 4 | 482 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
