Assay Details
| Gene Symbol: atp6ap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173265.2 | NP_775372.2 | 3 - 4 | 347 | 85 | |
| GenBank mRNA | AY099513.1 | - | 3 - 4 | 337 | 85 | |
| BC085541.1 | - | 3 - 4 | 347 | 85 | ||
| BC164542.1 | - | 3 - 4 | 359 | 85 | ||
More Information
Set Membership: |
|

atp6ap1b
Genomic Map