Assay Details
| Gene Symbol: tph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178306.3 | NP_840091.2 | 4 - 5 | 571 | 103 | |
| XM_017354058.2 | XP_017209547.1 | 4 - 5 | 547 | 103 | ||
| GenBank mRNA | AF548566.1 | - | 4 - 5 | 583 | 103 | |
| BC059550.1 | - | 4 - 5 | 572 | 103 | ||
More Information
Set Membership: |
|

tph1a
Genomic Map