Assay Details
| Gene Symbol: cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194408.2 | NP_919389.2 | 12 - 13 | 1600 | 66 | |
| XM_005162678.4 | XP_005162735.1 | 12 - 13 | 1610 | 66 | ||
| GenBank mRNA | AY007219.1 | - | 12 - 13 | 1304 | 66 | |
| BC045878.1 | - | 12 - 13 | 1494 | 66 | ||
| BC153307.1 | - | 12 - 13 | 1525 | 66 | ||
More Information
Set Membership: |
|

cdk8
Genomic Map