Assay Details
| Gene Symbol: gad1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194419.1 | NP_919400.1 | 12 - 13 | 1321 | 92 | |
| XM_009302103.3 | XP_009300378.1 | 12 - 13 | 1313 | 92 | ||
| XM_009302104.3 | XP_009300379.1 | 11 - 12 | 1498 | 92 | ||
| GenBank mRNA | AB183390.1 | - | 12 - 13 | 1505 | 92 | |
| AF017266.1 | - | 6 - 7 | 542 | 92 | ||
| BC047851.1 | - | 12 - 13 | 1321 | 92 | ||
More Information
Set Membership: |
|

gad1b
Genomic Map