Assay Details
| Gene Symbol: gad1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194419.1 | NP_919400.1 | 13 - 14 | 1405 | 69 | |
| XM_009302103.3 | XP_009300378.1 | 13 - 14 | 69 | |||
| XM_009302104.3 | XP_009300379.1 | 12 - 13 | 69 | |||
| GenBank mRNA | AB183390.1 | - | 13 - 14 | 69 | ||
| AF017266.1 | - | 7 - 8 | 69 | |||
| BC047851.1 | - | 13 - 14 | 1405 | 69 | ||
More Information
Set Membership: |
|

gad1b
Genomic Map