Assay Details
| Gene Symbol: gad1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194419.1 | NP_919400.1 | 7 - 8 | 892 | 64 | |
| XM_009302103.3 | XP_009300378.1 | 7 - 8 | 884 | 64 | ||
| XM_009302104.3 | XP_009300379.1 | 6 - 7 | 1069 | 64 | ||
| GenBank mRNA | AB183390.1 | - | 7 - 8 | 1076 | 64 | |
| AF017266.1 | - | 1 - 2 | 113 | 64 | ||
| AF042374.1 | - | 1 - 2 | 102 | 64 | ||
| BC047851.1 | - | 7 - 8 | 892 | 64 | ||
More Information
Set Membership: |
|

gad1b
Genomic Map