Assay Details
| Gene Symbol: gad1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194419.1 | NP_919400.1 | 9 - 10 | 1092 | 109 | |
| XM_009302103.3 | XP_009300378.1 | 9 - 10 | 1084 | 109 | ||
| XM_009302104.3 | XP_009300379.1 | 8 - 9 | 1269 | 109 | ||
| GenBank mRNA | AB183390.1 | - | 9 - 10 | 1276 | 109 | |
| AF017266.1 | - | 3 - 4 | 313 | 109 | ||
| AF042374.1 | - | 3 - 4 | 302 | 109 | ||
| BC047851.1 | - | 9 - 10 | 1092 | 109 | ||
More Information
Set Membership: |
|

gad1b
Genomic Map