Assay Details
| Gene Symbol: gspt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198806.1 | NP_942101.1 | 2 - 3 | 387 | 65 | |
| GenBank mRNA | BC053244.1 | - | 2 - 3 | 387 | 65 | |
More Information
Set Membership: |
|

gspt1l
Genomic Map