Assay Details
| Gene Symbol: ivns1abpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199279.2 | NP_954973.2 | 4 - 5 | 721 | 74 | |
| XM_021468374.1 | XP_021324049.1 | 4 - 5 | 660 | 74 | ||
| GenBank mRNA | BC046068.1 | - | 5 - 6 | 626 | 74 | |
| BC066513.1 | - | 4 - 5 | 689 | 74 | ||
| DN860974.1 | - | 2 - 3 | 287 | 74 | ||
More Information
Set Membership: |
|

ivns1abpa
Genomic Map