Assay Details
| Gene Symbol: ivns1abpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199279.2 | NP_954973.2 | 7 - 8 | 1106 | 60 | |
| XM_021468374.1 | XP_021324049.1 | 7 - 8 | 1045 | 60 | ||
| GenBank mRNA | BC046068.1 | - | 8 - 9 | 1011 | 60 | |
| BC066513.1 | - | 7 - 8 | 1074 | 60 | ||
| DN860974.1 | - | 5 - 6 | 672 | 60 | ||
More Information
Set Membership: |
|

ivns1abpa
Genomic Map