Assay Details
| Gene Symbol: atad5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082829.2 | NP_001076298.2 | 4 - 5 | 1389 | 149 | |
| XM_005166177.4 | XP_005166234.1 | 4 - 5 | 1454 | 149 | ||
| GenBank mRNA | BC057535.1 | - | 4 - 5 | 1389 | 149 | |
More Information
Set Membership: |
|

atad5b
Genomic Map