Assay Details
| Gene Symbol: atad5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082829.2 | NP_001076298.2 | 6 - 7 | 1694 | 118 | |
| GenBank mRNA | BC057535.1 | - | 6 - 7 | 1694 | 118 | |
More Information
Set Membership: |
|

atad5b
Genomic Map