Assay Details
| Gene Symbol: atp6v0b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199561.2 | NP_955855.2 | 3 - 4 | 393 | 62 | |
| GenBank mRNA | BC058877.1 | - | 3 - 4 | 381 | 62 | |
| EH446151.1 | - | 3 - 4 | 395 | 62 | ||
More Information
Set Membership: |
|

atp6v0b
Genomic Map