Assay Details
| Gene Symbol: csrnp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199619.1 | NP_955913.1 | 4 - 5 | 821 | 65 | |
| XM_005162674.4 | XP_005162731.1 | 4 - 5 | 950 | 65 | ||
| GenBank mRNA | BC057518.1 | - | 4 - 5 | 821 | 65 | |
More Information
Set Membership: |
|

csrnp1b
Genomic Map