Assay Details
| Gene Symbol: csrnp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199619.1 | NP_955913.1 | 5 - 6 | 1355 | 69 | |
| GenBank mRNA | BC057518.1 | - | 5 - 6 | 1355 | 69 | |
More Information
Set Membership: |
|

csrnp1b
Genomic Map