Assay Details
| Gene Symbol: snrpd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199760.1 | NP_956054.1 | 2 - 3 | 245 | 71 | |
| GenBank mRNA | BC045929.1 | - | 2 - 3 | 245 | 71 | |
| BC165492.1 | - | 1 - 2 | 154 | 71 | ||
More Information
Set Membership: |
|

snrpd3l
Genomic Map