Assay Details
| Gene Symbol: rcbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199991.1 | NP_956285.1 | 6 - 7 | 842 | 66 | |
| XM_005170970.4 | XP_005171027.1 | 6 - 7 | 871 | 66 | ||
| GenBank mRNA | BC045877.1 | - | 6 - 7 | 842 | 66 | |
| BC165711.1 | - | 5 - 6 | 752 | 66 | ||
More Information
Set Membership: |
|

rcbtb1
Genomic Map