Assay Details
| Gene Symbol: rab11al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200123.1 | NP_956417.1 | 3 - 4 | 788 | 70 | |
| GenBank mRNA | BC055141.1 | - | 3 - 4 | 788 | 70 | |
More Information
Set Membership: |
|

rab11al
Genomic Map