Assay Details
| Gene Symbol: rpap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200204.2 | NP_956498.2 | 15 - 16 | 1732 | 67 | |
| GenBank mRNA | BC045972.1 | - | 15 - 16 | 1664 | 67 | |
More Information
Set Membership: |
|

rpap3
Genomic Map