Assay Details
| Gene Symbol: ildr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200393.1 | NP_956687.1 | 6 - 7 | 913 | 76 | |
| GenBank mRNA | BC053314.1 | - | 6 - 7 | 913 | 76 | |
| CU458974.1 | - | 6 - 7 | 920 | 76 | ||
More Information
Set Membership: |
|

ildr1a
Genomic Map