Assay Details
| Gene Symbol: tmem218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200398.2 | NP_956692.1 | 3 - 4 | 362 | 59 | |
| XM_021479118.1 | XP_021334793.1 | 2 - 3 | 516 | 59 | ||
| XM_021479119.1 | XP_021334794.1 | 3 - 4 | 265 | 59 | ||
| GenBank mRNA | BC054131.1 | - | 3 - 4 | 363 | 59 | |
More Information
Set Membership: |
|

tmem218
Genomic Map