Assay Details
| Gene Symbol: adam10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200420.1 | NP_956714.1 | 1251 | 85 | ||
| XM_021470558.1 | XP_021326233.1 | 9 - 10 | 1309 | 85 | ||
| GenBank mRNA | BC054638.1 | - | 1251 | 85 | ||
| BC161665.1 | - | 1251 | 85 | |||
More Information
Set Membership: |
|

adam10b
Genomic Map