Assay Details
| Gene Symbol: polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200518.2 | NP_956812.2 | 9 - 10 | 1660 | 59 | |
| GenBank mRNA | BC097040.1 | - | 9 - 10 | 1660 | 59 | |
| BC165683.1 | - | 9 - 10 | 1645 | 59 | ||
More Information
Set Membership: |
|

polr1b
Genomic Map