Assay Details
| Gene Symbol: crnkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200946.1 | NP_957240.1 | 4 - 5 | 507 | 58 | |
| GenBank mRNA | BC044369.1 | - | 4 - 5 | 507 | 58 | |
More Information
Set Membership: |
|

crnkl1
Genomic Map