Assay Details
| Gene Symbol: zfand5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200949.1 | NP_957243.1 | 4 - 5 | 623 | 72 | |
| GenBank mRNA | BC050491.1 | - | 4 - 5 | 623 | 72 | |
| BC165024.1 | - | 3 - 4 | 524 | 72 | ||
More Information
Set Membership: |
|

zfand5b
Genomic Map