Assay Details
| Gene Symbol: atp2a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200965.1 | NP_957259.1 | 17 - 18 | 2730 | 90 | |
| XM_005166798.4 | XP_005166855.1 | 17 - 18 | 2756 | 90 | ||
| XM_005166799.4 | XP_005166856.1 | 17 - 18 | 2752 | 90 | ||
| XM_021478220.1 | XP_021333895.1 | 17 - 18 | 2756 | 90 | ||
| GenBank mRNA | BC045327.1 | - | 17 - 18 | 2730 | 90 | |
| BC165245.1 | - | 16 - 17 | 2549 | 90 | ||
More Information
Set Membership: |
|

atp2a2a
Genomic Map