Assay Details
| Gene Symbol: prkcbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200978.1 | NP_957272.1 | 4 - 5 | 539 | 71 | |
| XM_005164005.4 | XP_005164062.1 | 4 - 5 | 566 | 71 | ||
| GenBank mRNA | AY130432.1 | - | 2 - 3 | 191 | 71 | |
| BC055154.1 | - | 4 - 5 | 539 | 71 | ||
More Information
Set Membership: |
|

prkcbb
Genomic Map