Assay Details
| Gene Symbol: arf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201504.1 | NP_958912.1 | 3 - 4 | 326 | 82 | |
| GenBank mRNA | BC050487.1 | - | 3 - 4 | 326 | 82 | |
| BC066632.1 | - | 3 - 4 | 298 | 82 | ||
More Information
Set Membership: |
|

arf2a
Genomic Map