Assay Details
| Gene Symbol: slc3a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201514.2 | NP_958922.2 | 8 - 9 | 1275 | 117 | |
| XM_005161337.3 | XP_005161394.1 | 8 - 9 | 1310 | 117 | ||
| GenBank mRNA | BC044497.1 | - | 8 - 9 | 117 | ||
| BC063982.1 | - | 8 - 9 | 1275 | 117 | ||
More Information
Set Membership: |
|

slc3a2b
Genomic Map