Assay Details
| Gene Symbol: helt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207065.2 | NP_996948.1 | 1 - 2 | 222 | 81 | |
| GenBank mRNA | AY549496.1 | - | 1 - 2 | 176 | 81 | |
| BC092667.1 | - | 1 - 2 | 222 | 81 | ||
| BC164618.1 | - | 1 - 2 | 210 | 81 | ||
More Information
Set Membership: |
|

helt
Genomic Map