Assay Details
| Gene Symbol: dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212565.1 | NP_997730.1 | 3 - 4 | 822 | 60 | |
| GenBank mRNA | AY732218.2 | - | 3 - 4 | 647 | 60 | |
| BC059592.1 | - | 3 - 4 | 822 | 60 | ||
More Information
Set Membership: |
|

dusp5
Genomic Map