Assay Details
| Gene Symbol: ptpn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212654.1 | NP_997819.1 | 1 - 2 | 185 | 71 | |
| GenBank mRNA | BC044373.1 | - | 1 - 2 | 185 | 71 | |
| BC066698.1 | - | 1 - 2 | 178 | 71 | ||
| BC164031.1 | - | 1 - 2 | 89 | 71 | ||
More Information
Set Membership: |
|

ptpn2b
Genomic Map