Assay Details
| Gene Symbol: dnajb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212659.1 | NP_997824.1 | 5 - 6 | 766 | 61 | |
| GenBank mRNA | BC055389.1 | - | 5 - 6 | 766 | 61 | |
More Information
Set Membership: |
|

dnajb12a
Genomic Map