Assay Details
| Gene Symbol: eif2s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212675.1 | NP_997840.1 | 3 - 4 | 354 | 67 | |
| GenBank mRNA | BC066706.1 | - | 3 - 4 | 354 | 67 | |
More Information
Set Membership: |
|

eif2s2
Genomic Map