Assay Details
| Gene Symbol: hlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212701.1 | NP_997866.1 | 2 - 3 | 668 | 69 | |
| GenBank mRNA | BC066687.1 | - | 2 - 3 | 668 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hlx1| Gene Symbol: hlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212701.1 | NP_997866.1 | 2 - 3 | 668 | 69 | |
| GenBank mRNA | BC066687.1 | - | 2 - 3 | 668 | 69 | |
Set Membership: |
|