Assay Details
| Gene Symbol: zpax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040389.1 | NP_001035479.1 | 12 - 13 | 1687 | 144 | |
| NM_212718.1 | NP_997883.1 | 7 - 8 | 1083 | 144 | ||
| GenBank mRNA | AF509492.1 | - | 7 - 8 | 1083 | 144 | |
| BC115150.1 | - | 12 - 13 | 1687 | 144 | ||
| BC124757.1 | - | 9 - 10 | 1346 | 144 | ||
| BC129309.1 | - | 9 - 10 | 1372 | 144 | ||
| BC146721.1 | - | 9 - 10 | 1325 | 144 | ||
More Information
Set Membership: |
|

zpax1
Genomic Map