Assay Details
| Gene Symbol: zpax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040389.1 | NP_001035479.1 | 14 - 15 | 1999 | 80 | |
| NM_212718.1 | NP_997883.1 | 9 - 10 | 1395 | 80 | ||
| GenBank mRNA | AF509492.1 | - | 9 - 10 | 1395 | 80 | |
| BC115150.1 | - | 14 - 15 | 1999 | 80 | ||
| BC124071.1 | - | 1580 | 80 | |||
| BC124757.1 | - | 11 - 12 | 1658 | 80 | ||
| BC129309.1 | - | 11 - 12 | 1684 | 80 | ||
| BC146721.1 | - | 11 - 12 | 1637 | 80 | ||
More Information
Set Membership: |
|

zpax1
Genomic Map