Assay Details
| Gene Symbol: adka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272084.1 | NP_001259013.1 | 8 - 9 | 795 | 68 | |
| NM_212791.2 | NP_997956.1 | 8 - 9 | 869 | 68 | ||
| GenBank mRNA | BC044481.1 | - | 8 - 9 | 821 | 68 | |
| BC063961.1 | - | 8 - 9 | 788 | 68 | ||
| BC164029.1 | - | 7 - 8 | 719 | 68 | ||
More Information
Set Membership: |
|

adka
Genomic Map