Assay Details
| Gene Symbol: msi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320102.1 | NP_001307031.1 | 13 - 14 | 1180 | 77 | |
| XM_009305537.3 | XP_009303812.1 | 13 - 14 | 1294 | 77 | ||
| GenBank mRNA | BC045335.1 | - | 13 - 14 | 1161 | 77 | |
| EH471460.1 | - | 2 - 3 | 499 | 77 | ||
More Information
Set Membership: |
|

msi2a
Genomic Map