Assay Details
| Gene Symbol: msi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320102.1 | NP_001307031.1 | 14 - 15 | 1260 | 69 | |
| XM_009305537.3 | XP_009303812.1 | 14 - 15 | 1374 | 69 | ||
| GenBank mRNA | BC045335.1 | - | 14 - 15 | 1241 | 69 | |
| EH471460.1 | - | 1 - 2 | 419 | 69 | ||
More Information
Set Membership: |
|

msi2a
Genomic Map