Assay Details
| Gene Symbol: uts2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212848.3 | NP_998013.2 | 2 - 3 | 247 | 108 | |
| GenBank mRNA | AY305004.1 | - | 2 - 3 | 253 | 108 | |
| BC162257.1 | - | 2 - 3 | 247 | 108 | ||
| BC162265.1 | - | 2 - 3 | 247 | 108 | ||
More Information
Set Membership: |
|

uts2a
Genomic Map