Assay Details
| Gene Symbol: ren | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212860.1 | NP_998025.1 | 7 - 8 | 969 | 60 | |
| GenBank mRNA | AY167037.1 | - | 7 - 8 | 969 | 60 | |
| BC162339.1 | - | 7 - 8 | 969 | 60 | ||
| BC162351.1 | - | 7 - 8 | 969 | 60 | ||
More Information
Set Membership: |
|

ren
Genomic Map