Assay Details
| Gene Symbol: ddx39ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212977.2 | NP_998142.1 | 7 - 8 | 1103 | 94 | |
| GenBank mRNA | BC044169.1 | - | 7 - 8 | 1073 | 94 | |
| BC067555.1 | - | 7 - 8 | 1043 | 94 | ||
| BC128804.1 | - | 7 - 8 | 1048 | 94 | ||
| BC165847.1 | - | 6 - 7 | 1006 | 94 | ||
More Information
Set Membership: |
|

ddx39ab
Genomic Map