Assay Details
| Gene Symbol: ddx39ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212977.2 | NP_998142.1 | 8 - 9 | 1250 | 95 | |
| GenBank mRNA | BC044169.1 | - | 8 - 9 | 1220 | 95 | |
| BC067555.1 | - | 8 - 9 | 1190 | 95 | ||
| BC128804.1 | - | 8 - 9 | 1195 | 95 | ||
| BC165847.1 | - | 7 - 8 | 1153 | 95 | ||
More Information
Set Membership: |
|

ddx39ab
Genomic Map