Assay Details
| Gene Symbol: itpk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213017.1 | NP_998182.1 | 4 - 5 | 578 | 118 | |
| XM_009300123.3 | XP_009298398.1 | 4 - 5 | 599 | 118 | ||
| GenBank mRNA | BC050497.1 | - | 4 - 5 | 578 | 118 | |
| BC164805.1 | - | 3 - 4 | 278 | 118 | ||
More Information
Set Membership: |
|

itpk1a
Genomic Map