Assay Details
| Gene Symbol: tpm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024467.1 | NP_001019638.1 | 7 - 8 | 874 | 114 | |
| NM_213158.1 | NP_998323.1 | 6 - 7 | 976 | 114 | ||
| XM_017353110.1 | XP_017208599.1 | 7 - 8 | 917 | 114 | ||
| XM_017353111.2 | XP_017208600.1 | 6 - 7 | 984 | 114 | ||
| GenBank mRNA | BC053144.1 | - | 6 - 7 | 976 | 114 | |
| BC085518.1 | - | 7 - 8 | 874 | 114 | ||
More Information
Set Membership: |
|

tpm4a
Genomic Map